Tisk…

5'-Ethyl-2'-hydroxyacetophenone 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Ethyl-2-hydroxyacetophenone

OR928508-1G OR928508-5G
APOSOR928508-1GEA 1210 CZK
APOSOR928508-1G APOSOR928508-5G
5'-Ethyl-2'-hydroxyacetophenone 95%
5'-Ethyl-2'-hydroxyacetophenone
Vzorec: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00053308
CAS číslo: 24539-92-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR