Tisk…

2-Piperidin-1-ylmethylphenylamine 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Piperidin-1-ylmethyl-aniline

OR928481-1G OR928481-5G
APOSOR928481-1GEA 3540 CZK
APOSOR928481-1G APOSOR928481-5G
2-Piperidin-1-ylmethylphenylamine 97%
2-Piperidin-1-ylmethylphenylamine
Vzorec: C₁₂H₁₈N₂
MW: 190,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114503
CAS číslo: 19577-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR