Tisk…

1,2,3,4-Tetrahydro-2,6-naphthyridine hydrochloride 95%

Dodavatel: Apollo Scientific

OR928457-1G OR928457-250MG
APOSOR928457-1GEA 27910 CZK
APOSOR928457-1G APOSOR928457-250MG
1,2,3,4-Tetrahydro-2,6-naphthyridine hydrochloride 95%
1,2,3,4-Tetrahydro-2,6-naphthyridine hydrochloride
Vzorec: C₈H₁₁ClN₂
MW: 170,64 g/mol
Storage Temperature: Ambient
CAS číslo: 1416352-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR