Dibenzo[b,d]furan-4-carbaldehyde 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Dibenzofuran-4-carboxaldehyde
OR928449-5G
APOSOR928449-5GEA
2820
CZK
APOSOR928449-5G
Dibenzo[b,d]furan-4-carbaldehyde 96%
Dibenzo[b,d]furan-4-carbaldehyde
Vzorec:
C₁₃H₈O₂ MW: 196,21 g/mol Bod tání: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD03306027 CAS číslo: 96706-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...