Ethyl 3-(methylamino)-2-butenoate 95%
Dodavatel: Apollo Scientific
OR928436-100G
OR928436-25G
APOSOR928436-100GEA
4270
CZK
APOSOR928436-100G
APOSOR928436-25G
Ethyl 3-(methylamino)-2-butenoate 95%
Ethyl-3-(methylamino)-2-butenoate
Vzorec:
C₇H₁₃NO₂ MW: 143.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03060877 CAS číslo: 870-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...