4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid 98%
Dodavatel: Apollo Scientific
OR928406-1G
OR928406-250MG
APOSOR928406-1GEA
17990
CZK
APOSOR928406-1G
APOSOR928406-250MG
4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid 98%
4,7-Dihydro-5H-thieno[2,3-c]pyran-3-carboxylic acid
Vzorec:
C₈H₈O₃S Storage Temperature: Ambient |
MDL Number:
MFCD17016073 CAS číslo: 1169491-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...