Tisk…

1-(1-Methyl-1H-imidazol-2-yl)ethanone 95%

Dodavatel: Apollo Scientific
Varování

OR928251-1G OR928251-250MG OR928251-5G
APOSOR928251-1GEA 3770 CZK
APOSOR928251-1G APOSOR928251-250MG APOSOR928251-5G
1-(1-Methyl-1H-imidazol-2-yl)ethanone 95%
1-(1-Methyl-1H-imidazol-2-yl)ethanone
Vzorec: C₆H₈N₂O
MW: 124,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08668178
CAS číslo: 85692-37-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR