Tisk…

1-(Trimethylsilyloxy)cyclopentene 95%

Dodavatel: Apollo Scientific

Synonyms: (Cyclopent-1-en-1-yloxy)trimethylsilane, (1-Cyclopentenyloxy)trimethylsilane

OR928213-5G
APOSOR928213-5GEA 2820 CZK
APOSOR928213-5G
1-(Trimethylsilyloxy)cyclopentene 95%
1-(Trimethylsilyloxy)cyclopentene
Vzorec: C₈H₁₆OSi
MW: 156,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001395
CAS číslo: 19980-43-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR