Tisk…

4-(Dimethylamino)benzamide 95%

Dodavatel: Apollo Scientific

OR928188-1G OR928188-5G
APOSOR928188-1GEA 2440 CZK
APOSOR928188-1G APOSOR928188-5G
4-(Dimethylamino)benzamide 95%
4-(Dimethylamino)benzamide
Vzorec: C₉H₁₂N₂O
MW: 164,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01216782
CAS číslo: 6083-47-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR