Tisk…

4-Ethoxy-3-(hydroxymethyl)benzaldehyde 95%

Dodavatel: Apollo Scientific

OR928147-1G OR928147-5G
APOSOR928147-1GEA 4770 CZK
APOSOR928147-1G APOSOR928147-5G
4-Ethoxy-3-(hydroxymethyl)benzaldehyde 95%
4-Ethoxy-3-(hydroxymethyl)benzaldehyde
Vzorec: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD03180283
CAS číslo: 917746-81-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR