Tisk…

2-(5-Formyl-2-furyl)benzonitrile 95%

Dodavatel: Apollo Scientific

OR928056-1G OR928056-5G
APOSOR928056-1GEA 4770 CZK
APOSOR928056-1G APOSOR928056-5G
2-(5-Formyl-2-furyl)benzonitrile 95%
2-(5-Formyl-2-furyl)benzonitrile
Vzorec: C₁₂H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD02630467
CAS číslo: 299442-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR