Di(2-pyridyl) ketone 97+%
Dodavatel: Apollo Scientific
Synonyms:
Di(pyridin-2-yl)methanone
OR928006-100G
OR928006-25G
APOSOR928006-100GEA
11180
CZK
APOSOR928006-100G
APOSOR928006-25G
Di(2-pyridyl) ketone 97+%
Di(2-pyridyl) ketone
Vzorec:
C₁₁H₈N₂O MW: 184,20 g/mol Bod tání: 52…55 °C Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006288 CAS číslo: 19437-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...