Tisk…

2-Chloropyrazolo[1,5-a]pyridine 95%

Dodavatel: Apollo Scientific

OR927933-1G OR927933-250MG
APOSOR927933-1GEA 26450 CZK
APOSOR927933-1G APOSOR927933-250MG
2-Chloropyrazolo[1,5-a]pyridine 95%
2-Chloropyrazolo[1,5-a]pyridine
Vzorec: C₇H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06739221
CAS číslo: 60637-33-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR