4-Carboxyphenylacetic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Carboxyphenyl)acetic acid, 4-(Carboxymethyl)benzoic acid
OR927867-100G
OR927867-25G
OR927867-5G
APOSOR927867-100GEA
7010
CZK
APOSOR927867-100G
APOSOR927867-25G
APOSOR927867-5G
4-Carboxyphenylacetic acid 99%
4-Carboxyphenylacetic acid
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01074834 CAS číslo: 501-89-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...