Tisk…

Dipropyl sulphone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(Propylsulphonyl)propane

OR927856-25G
APOSOR927856-25GEA 5560 CZK
APOSOR927856-25G
Dipropyl sulphone 95%
Dipropyl sulphone
Vzorec: C₆H₁₄O₂S
MW: 150,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007570
CAS číslo: 598-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR