Tisk…

3-(1,3-Dioxolan-2-yl)aniline 95%

Dodavatel: Apollo Scientific

OR927763-1G
APOSOR927763-1GEA 1490 CZK
APOSOR927763-1G
3-(1,3-Dioxolan-2-yl)aniline 95%
3-(1,3-Dioxolan-2-yl)aniline
Vzorec: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00272236
CAS číslo: 6398-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR