2'-Amino-4',5'-dimethoxyacetophenone 96%
Dodavatel: Apollo Scientific
Synonyms:
1-(2-Amino-4,5-dimethoxyphenyl)ethanone
OR927720-25G
APOSOR927720-25GEA
4800
CZK
APOSOR927720-25G
2'-Amino-4',5'-dimethoxyacetophenone 96%
2'-Amino-4',5'-dimethoxyacetophenone
Vzorec:
C₁₀H₁₃NO₃ MW: 195,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016646 CAS číslo: 4101-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...