3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
Dodavatel: Apollo Scientific
|
Varování
|
OR927621-100MG
APOSOR927621-100MGEA
2030
CZK
APOSOR927621-100MG
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
3,4-Dihydro-2H-pyrano[2,3-b]pyridin-6-amine
Vzorec:
C₈H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD20487103 CAS číslo: 1346447-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...