Tisk…

(1-Methyl-1H-imidazol-2-yl)methylamine 98%

Dodavatel: Apollo Scientific

Synonyms: (1-Methyl-1H-imidazol-2-yl)methanamine

OR927555-1G OR927555-5G
APOSOR927555-1GEA 1620 CZK
APOSOR927555-1G APOSOR927555-5G
(1-Methyl-1H-imidazol-2-yl)methylamine 98%
(1-Methyl-1H-imidazol-2-yl)methylamine
Vzorec: C₅H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD06660876
CAS číslo: 124312-73-8
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR