Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 6-bromo-1-methylindole-2-carboxylate
OR927482-1G
OR927482-5G
APOSOR927482-1GEA
7250
CZK
APOSOR927482-1G
APOSOR927482-5G
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate 95%
Methyl-6-bromo-1-methyl-1H-indole-2-carboxylate
Vzorec:
C₁₁H₁₀BrNO₂ MW: 268,11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07364734 CAS číslo: 680569-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...