Tisk…

1-(Pyridin-2-yl)cyclopropanecarboxylic acid 96%

Dodavatel: Apollo Scientific

OR927460-1G OR927460-250MG OR927460-5G
APOSOR927460-1GEA 5230 CZK
APOSOR927460-1G APOSOR927460-250MG APOSOR927460-5G
1-(Pyridin-2-yl)cyclopropanecarboxylic acid 96%
1-(Pyridin-2-yl)cyclopropanecarboxylic acid
Vzorec: C₉H₉NO₂
MW: 163.1733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864786
CAS číslo: 162960-26-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR