Tisk…

Fmoc-N-Me-Nle-OH 97%

Dodavatel: Apollo Scientific

Synonyms: N-Fmoc-N-methyl-L-norleucine

OR927345-25G
APOSOR927345-25GEA 3970 CZK
APOSOR927345-25G
Fmoc-N-Me-Nle-OH 97%
Fmoc-N-Me-Nle-OH
Vzorec: C₂₂H₂₅NO₄
MW: 367,445 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235878
CAS číslo: 112883-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR