Tisk…

8-Methylimidazo[1,2-a]pyridine-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR927287-1G OR927287-250MG
APOSOR927287-1GEA 5530 CZK
APOSOR927287-1G APOSOR927287-250MG
8-Methylimidazo[1,2-a]pyridine-2-carbaldehyde 95%
8-Methylimidazo[1,2-a]pyridine-2-carbaldehyde
Vzorec: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD06739274
CAS číslo: 143982-39-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR