Tisk…

1H-Pyrrolo[2,3-b]pyridine-6-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR927275-1G OR927275-250MG
APOSOR927275-1GEA 4960 CZK
APOSOR927275-1G APOSOR927275-250MG
1H-Pyrrolo[2,3-b]pyridine-6-carboxylic acid 95%
1H-Pyrrolo[2,3-b]pyridine-6-carboxylic acid
Vzorec: C₈H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08272223
CAS číslo: 898746-35-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR