1-Methylindazole-5-carboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Methyl-1H-indazole-5-carboxylic acid
OR927241-1G
OR927241-250MG
OR927241-5G
APOSOR927241-1GEA
1040
CZK
APOSOR927241-1G
APOSOR927241-250MG
APOSOR927241-5G
1-Methylindazole-5-carboxylic acid 96%
1-Methylindazole-5-carboxylic acid
Vzorec:
C₉H₈N₂O₂ MW: 176,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11901714 CAS číslo: 1176754-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...