(4-Ethoxyphenyl)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR927167-100G
OR927167-25G
OR927167-5G
APOSOR927167-100GEA
3230
CZK
APOSOR927167-100G
APOSOR927167-25G
APOSOR927167-5G
(4-Ethoxyphenyl)acetic acid 98%
(4-Ethoxyphenyl)acetic acid
Vzorec:
(C₆H₄(OCH₂CH₃))CH₂COOH Bod tání: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004346 CAS číslo: 4919-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...