Tisk…

4-(1H-Imdazol-4-yl)piperidine 96%

Dodavatel: Apollo Scientific
Varování

OR927063-1G OR927063-250MG OR927063-5G
APOSOR927063-1GEA 10520 CZK
APOSOR927063-1G APOSOR927063-250MG APOSOR927063-5G
4-(1H-Imdazol-4-yl)piperidine 96%
4-(1H-Imdazol-4-yl)piperidine
Vzorec: C₈H₁₃N₃
Storage Temperature: Refrigerator
CAS číslo: 106243-23-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR