5-Amino-4-bromopyrimidine 95%
Dodavatel: Apollo Scientific
OR927026-1G
OR927026-250MG
APOSOR927026-1GEA
5530
CZK
APOSOR927026-1G
APOSOR927026-250MG
5-Amino-4-bromopyrimidine 95%
5-Amino-4-bromopyrimidine
Vzorec:
C₄H₄BrN₃ Storage Temperature: Ambient |
MDL Number:
MFCD09999203 CAS číslo: 849353-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...