1-Methylpyrrolo[2,3-b]pyridin-5-amine 97%
Dodavatel: Apollo Scientific
OR926884-1G
OR926884-250MG
APOSOR926884-1GEA
18420
CZK
APOSOR926884-1G
APOSOR926884-250MG
1-Methylpyrrolo[2,3-b]pyridin-5-amine 97%
1-Methylpyrrolo[2,3-b]pyridin-5-amine
Vzorec:
C₈H₉N₃ MW: 147,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12923911 CAS číslo: 883986-76-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...