Benzo[d][1,3]dioxole-4-carbonyl chloride 95%
Dodavatel: Apollo Scientific
OR926814-1G
OR926814-5G
APOSOR926814-1GEA
9720
CZK
APOSOR926814-1G
APOSOR926814-5G
Benzo[d][1,3]dioxole-4-carbonyl chloride 95%
Benzo[d][1,3]dioxole-4-carbonyl chloride
Vzorec:
C₈H₅ClO₃ Storage Temperature: Ambient |
CAS číslo:
66411-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...