5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR926791-100MG
OR926791-1G
OR926791-250MG
APOSOR926791-100MGEA
3310
CZK
APOSOR926791-100MG
APOSOR926791-1G
APOSOR926791-250MG
5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 95%
5-Methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid
Vzorec:
C₉H₈N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD08448208 CAS číslo: 17288-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...