Dimethyl isopropylmalonate 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR926639-25G
OR926639-100G
APOSOR926639-25GEA
2690
CZK
APOSOR926639-25G
APOSOR926639-100G
Dimethyl isopropylmalonate 98%
Dimethyl isopropylmalonate
Vzorec:
C₈H₁₄O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00236605 CAS číslo: 51122-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...