N,N-Diethylnicotinamide 97%
Dodavatel: Apollo Scientific
Synonyms:
Nicotinic Acid Diethylamide
OR926552-100G
OR926552-500G
APOSOR926552-100GEA
4770
CZK
APOSOR926552-100G
APOSOR926552-500G
N,N-Diethylnicotinamide 97%
N,N-Diethylnicotinamide
Vzorec:
C₁₀H₁₄N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00006386 CAS číslo: 59-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...