Tisk…

Boc-3-bromo-D-phenylalanine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Boc-3-bromo-D-phenylalanine, (R)-3-(3-Bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

OR926518-5G
APOSOR926518-5GEA 4600 CZK
APOSOR926518-5G
Boc-3-bromo-D-phenylalanine 97%
Boc-3-bromo-D-phenylalanine
Vzorec: C₁₄H₁₈BrNO₄
MW: 344,205 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01317704
CAS číslo: 261360-77-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR