Tisk…

1-Ethyl-1H-indole-4-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR926423-1G OR926423-250MG
APOSOR926423-1GEA 10750 CZK
APOSOR926423-1G APOSOR926423-250MG
1-Ethyl-1H-indole-4-carbaldehyde 95%
1-Ethyl-1H-indole-4-carbaldehyde
Vzorec: C₁₁H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD09455239
CAS číslo: 894852-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR