Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate 98%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 4-methyltetrahydro-2H-pyran-4-carboxylate
OR926394-1G
OR926394-5G
APOSOR926394-1GEA
2940
CZK
APOSOR926394-1G
APOSOR926394-5G
Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate 98%
Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate
Vzorec:
C₈H₁₄O₃ MW: 158,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12912101 CAS číslo: 443912-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...