Tisk…

tert-Butyl (cyanomethyl)carbamate 97%

Dodavatel: Apollo Scientific

Synonyms: N-(tert-Butoxycarbonyl)-2-aminoacetonitrile

OR926356-100G
APOSOR926356-100GEA 3970 CZK
APOSOR926356-100G
tert-Butyl (cyanomethyl)carbamate 97%
tert-Butyl (cyanomethyl)carbamate
Vzorec: C₇H₁₂N₂O₂
MW: 156.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239390
CAS číslo: 85363-04-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR