Tisk…

2,7-Dihydroxynaphthalene 98%

Dodavatel: Apollo Scientific

Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol

OR926339-1KG
APOSOR926339-1KGEA 0 CZK
APOSOR926339-1KG
2,7-Dihydroxynaphthalene 98%
2,7-Dihydroxynaphthalene
Vzorec: C₁₀H₈O₂
MW: 160,17 g/mol
Bod tání: 184…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS číslo: 582-17-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR