3-Chloroquinoline 98%
Dodavatel: Apollo Scientific
OR926222-1G
OR926222-5G
APOSOR926222-1GEA
0
CZK
APOSOR926222-1G
APOSOR926222-5G
3-Chloroquinoline 98%
3-Chloroquinoline
Vzorec:
C9H6ClN Storage Temperature: Ambient |
MDL Number:
MFCD00234489 CAS číslo: 612-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...