Tisk…

Ethyl-4-bromobenzoyl formate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-(4-bromophenyl)-2-oxoacetate

OR926113-5G
APOSOR926113-5GEA 1700 CZK
APOSOR926113-5G
Ethyl-4-bromobenzoyl formate 95%
Ethyl-4-bromobenzoyl formate
Vzorec: C₁₀H₉BrO₃
MW: 257,09 g/mol
Bod varu: 120…122 °C/0,1mmHg
Hustota: 1,438 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06659764
CAS číslo: 20201-26-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR