Tisk…

4,4-Dimethyl-3-oxopentanal 95%

Dodavatel: Apollo Scientific

OR926089-1G OR926089-25G OR926089-5G
APOSOR926089-1GEA 2900 CZK
APOSOR926089-1G APOSOR926089-25G APOSOR926089-5G
4,4-Dimethyl-3-oxopentanal 95%
4,4-Dimethyl-3-oxopentanal
Vzorec: C₇H₁₂O₂
MW: 128,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042806
CAS číslo: 23459-13-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR