3-Bromo-7-methylimidazo[1,2-a]pyrimidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR926040-100MG
OR926040-250MG
APOSOR926040-100MGEA
1660
CZK
APOSOR926040-100MG
APOSOR926040-250MG
3-Bromo-7-methylimidazo[1,2-a]pyrimidine 97%
3-Bromo-7-methylimidazo[1,2-a]pyrimidine
Vzorec:
C7H6BrN3 Storage Temperature: Refrigerator |
MDL Number:
MFCD08234998 CAS číslo: 375857-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...