Tisk…

N,N-Diethylsalicylamide 98%

Dodavatel: Apollo Scientific

OR925736-1G OR925736-5G
APOSOR925736-1GEA 3540 CZK
APOSOR925736-1G APOSOR925736-5G
N,N-Diethylsalicylamide 98%
N,N-Diethylsalicylamide
Vzorec: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00010816
CAS číslo: 19311-91-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR