Tisk…

1-Bicyclo[2.2.1]hept-2-ylethanamine 95%

Dodavatel: Apollo Scientific

OR925721-100MG OR925721-1G OR925721-250MG
APOSOR925721-100MGEA 4570 CZK
APOSOR925721-100MG APOSOR925721-1G APOSOR925721-250MG
1-Bicyclo[2.2.1]hept-2-ylethanamine 95%
1-Bicyclo[2.2.1]hept-2-ylethanamine
Vzorec: C₉H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD04116067
CAS číslo: 24520-60-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR