Tisk…

4-Chlorobutyl acetate 98%

Dodavatel: Apollo Scientific

OR925717-100G OR925717-500G
APOSOR925717-100GEA 857 CZK
APOSOR925717-100G APOSOR925717-500G
4-Chlorobutyl acetate 98%
4-Chlorobutyl acetate
Vzorec: C₆H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00001013
CAS číslo: 6962-92-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR