5-Cyclopropyl-isoxazole-3-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR925637-1G
OR925637-5G
APOSOR925637-1GEA
1740
CZK
APOSOR925637-1G
APOSOR925637-5G
5-Cyclopropyl-isoxazole-3-carboxylic acid 95%
5-Cyclopropyl-isoxazole-3-carboxylic acid
Vzorec:
C₇H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD05668325 CAS číslo: 110256-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...