Methyl-4-chloro-4-oxobutanoate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl succinyl chloride, Succinic acid monomethyl ester chloride, Methyl 4-Chloro-4-oxobutyrate, Methyl Succinyl chloride, 4-Chloro-4-oxobutyric acid methyll ester
OR925630-100G
OR925630-25G
APOSOR925630-100GEA
10190
CZK
APOSOR925630-100G
APOSOR925630-25G
Methyl-4-chloro-4-oxobutanoate 95%
Methyl-4-chloro-4-oxobutanoate
Vzorec:
C₅H₇ClO₃ MW: 150.56 g/mol Bod varu: 58…65 °C/3mmHg Hustota: 1.232 g/cm³ (25 °C) Bod vzplanutí: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000750 CAS číslo: 1490-25-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...