Chlorocarbonylsulphenyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Chlorothioformyl Chloride, Chloroformylsulphenyl Chloride
OR925562-10G
OR925562-25G
APOSOR925562-10GEA
2070
CZK
APOSOR925562-10G
APOSOR925562-25G
Chlorocarbonylsulphenyl chloride 95%
Chlorocarbonylsulphenyl chloride
Vzorec:
CCl₂OS Bod varu: 98 °C Hustota: 1,552 g/cm³ (25 °C) Bod vzplanutí: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000703 CAS číslo: 2757-23-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...