Methyl(5-phenyl[1,3,4]oxadiazol-2-ylmethyl)amine hydrochloride 95%
Dodavatel: Apollo Scientific
OR925519-1G
OR925519-250MG
OR925519-5G
APOSOR925519-1GEA
6220
CZK
APOSOR925519-1G
APOSOR925519-250MG
APOSOR925519-5G
Methyl(5-phenyl[1,3,4]oxadiazol-2-ylmethyl)amine hydrochloride 95%
Methyl(5-phenyl[1,3,4]oxadiazol-2-ylmethyl)amine hydrochloride
Vzorec:
C₁₀H₁₂ClN₃O Storage Temperature: Ambient |
MDL Number:
MFCD06738853 CAS číslo: 880361-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...