Tisk…

Fmoc-3,4-dehydroPro-OH 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2,5-dihydro-1H-pyrrole-2-carboxylic acid, Fmoc-3,4-dehydro-Pro-OH

OR925492-1G OR925492-250MG OR925492-500MG
APOSOR925492-1GEA 15310 CZK
APOSOR925492-1G APOSOR925492-250MG APOSOR925492-500MG
Fmoc-3,4-dehydroPro-OH 97%
Fmoc-3,4-dehydroPro-OH
Vzorec: C₂₀H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00151940
CAS číslo: 135837-63-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR